Learn More
Medchemexpress LLC 1-[(1-cyanocyclopropanecarbonyl)amino]-4-hydroxy-N-[(4-thiazolyl)benzyl]pyrrolidine carboxamide | 2097381-85-4 | MFCD30742947 | 99.8% | 523.65 | C27H33N5O4S | 1 ML
SDP
Catalog No. 5000212420
Encompass_Preferred
VH-298 is a potent, cell-permeable small-molecule inhibitor of the VHL:HIF-α protein-protein interaction, used to stabilize hypoxia-inducible factor α in biochemical and cellular studies. Supplied as a solid or as a 10 mM solution in DMSO.
Catalog No. 50-002-12420 Supplier Medchemexpress LLC Supplier No. HY10094710MM 1ML
Add to Cart
Add to Cart
VH-298 is a potent, cell-permeable small-molecule inhibitor of the VHL:HIF-α protein-protein interaction, used to stabilize hypoxia-inducible factor α in biochemical and cellular studies. Supplied as a solid or as a 10 mM solution in DMSO.