Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Cyclopentanediacetic Acid 98.0+%, TCI America™
SDP

Catalog No. C08385G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08385G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C08385G Supplier TCI America Supplier No. C08385G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16713-66-9
Molecular Formula C9H14O4
Molecular Weight (g/mol) 186.21
MDL Number MFCD00001388
InChI Key FWPVKDFOUXHOKQ-UHFFFAOYSA-N
Synonym 1,1-cyclopentanediacetic acid, 3,3-tetramethyleneglutaric acid, cyclopentane-1,1-diacetic acid, cyclopentanediacetic acid, tetramethyleneglutaric acid, tetramethylene glutaric acid, 2-1-carboxymethyl cyclopentyl acetic acid, 1,1-cyclopentanediaceticacid, chembl80679, 3,3'-tetramethylene glutaric acid
PubChem CID 85558
IUPAC Name 2-[1-(carboxymethyl)cyclopentyl]acetic acid
SMILES OC(=O)CC1(CC(O)=O)CCCC1

Specifications

Melting Point 177°C
Color White
Quantity 5 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Cyclopentanediacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.