Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Cyclopropanedicarboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C13095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13095G 5 g
C13091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13095G Supplier TCI America Supplier No. C13095G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 598-10-7
Molecular Formula C5H4O4
Molecular Weight (g/mol) 128.08
MDL Number MFCD00013727
InChI Key FDKLLWKMYAMLIF-UHFFFAOYSA-L
Synonym 1,1-cyclopropanedicarboxylic acid, 1,1-cyclopropanedicarboxylate, cyclopropane-1,1-dicarboxylicacid, cyclopropanedicarboxylic acid, vinaconic acid, cyclopropane, 3, vinyl-malonic acid, 9ty, ethylenemalonic acid, acmc-209mfz
PubChem CID 69017
IUPAC Name cyclopropane-1,1-dicarboxylate
SMILES [O-]C(=O)C1(CC1)C([O-])=O

Specifications

Melting Point 137°C
Color White
Quantity 5 g
UN Number 3261
Formula Weight 130.10
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Cyclopropanedicarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.