Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Dibromoferrocene 98.0+%, TCI America™
SDP

Catalog No. D28295G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28295G 5 g
D28291G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28295G Supplier TCI America Supplier No. D28295G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1293-65-8
Molecular Formula C10H8Br2Fe
Molecular Weight (g/mol) 343.83
MDL Number MFCD01075755
InChI Key LBOGSXRPHOKVET-UHFFFAOYSA-N
IUPAC Name 1,1'-Dibromoferrocene
SMILES [Fe].Brc1cccc1.Brc1cccc1

Specifications

Quantity 5 g
Formula Weight 343.83
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1′-Dibromoferrocene
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.