Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dichloro-2,2-bis(4-hydroxyphenyl)ethylene 98.0+%, TCI America™
SDP

Catalog No. D32235G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D32235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D32235G Supplier TCI America Supplier No. D32235G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14868-03-2
Molecular Formula C14H10Cl2O2
Molecular Weight (g/mol) 281.13
MDL Number MFCD00233304
InChI Key OWEYKIWAZBBXJK-UHFFFAOYSA-N
Synonym 1,1-Bis(4-hydroxyphenyl)-2,2-dichloroethylene, 4,4′C-(2,2-Dichlorovinylidene)bisphenol
PubChem CID 84677
ChEBI CHEBI:34030
IUPAC Name 4-[2,2-dichloro-1-(4-hydroxyphenyl)ethenyl]phenol
SMILES OC1=CC=C(C=C1)C(=C(Cl)Cl)C1=CC=C(O)C=C1

Specifications

Melting Point 216°C
Quantity 5 g
Formula Weight 281.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dichloro-2,2-bis(4-hydroxyphenyl)ethylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.