Learn More
Strem, An Ascensus Company CAS# 79-35-6. 25g. 1,1-Dichloro-2,2-difluoroethylene, min. 97%. MFCD00039338
SDP
Supplier:  Strem, An Ascensus Company 09236025G
Encompass
CAS# 79-35-6. 25g. 1,1-Dichloro-2,2-difluoroethylene, min. 97%. MFCD00039338. Molecular Weight: 133.00. Molecular Formula: CCl2=CF2. Color/form: colorless liq. Strem# 09-2360. http://www.strem.com/catalog/v/09-2360/
Catalog No. 50-901-09297