Learn More
Cambridge Isotope Laboratories DEUTERIUM HYDRIDE (D, 97%), 1 L, 13983-20-5
Catalog No. 501630916
Encompass
**This item is shipped in a Code K-460 stainless steel lecture bottle. The fee for the bottle will need added to the order as a separate charge per each. Please include part# NC1708343 to the order
Catalog No. 50-163-0916 Supplier Cambridge Isotope Laboratories Supplier No. DLM1941
**This item is shipped in a Code K-460 stainless steel lecture bottle. The fee for the bottle will need added to the order as a separate charge per each. Please include part# NC1708343 to the order