Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dichloropinacolin 98.0+%, TCI America™
SDP

Catalog No. D223325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D223325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D223325G Supplier TCI America Supplier No. D223325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22591-21-5
Molecular Formula C6H10Cl2O
Molecular Weight (g/mol) 169.05
MDL Number MFCD00069423
InChI Key UDWZXMQIEHAAQT-UHFFFAOYSA-N
Synonym tert-Butyl Dichloromethyl Ketone, 1,1-Dichloro-3,3-dimethyl-2-butanone, 1,1-Dichloropinacolone
PubChem CID 31432
IUPAC Name 1,1-dichloro-3,3-dimethylbutan-2-one
SMILES CC(C)(C)C(=O)C(Cl)Cl

Specifications

Melting Point 50°C
Color White
Boiling Point 178°C
Quantity 25 g
UN Number 2811
Formula Weight 169.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dichloropinacolin
RTECSNumber EL7500000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.