Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1'-Diethyl-2,2'-carbocyanine iodide, 96%

Catalog No. AAH31540MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH31540MD 250 mg
AAH3154003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH31540MD Supplier Thermo Scientific Chemicals Supplier No. H31540MD
  • $91.80
    $80.65 / Each of 1

    Save $11.15

    12% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 605-91-4 | C25H25IN2 | 480.39 g/mol

The fluorescence decay time of pinacyanol (1,1?-diethyl-2,2?-carbocyanine) iodide has been measured in the temperature range 95-225 K.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The fluorescence decay time of pinacyanol (1,1′-diethyl-2,2′-carbocyanine) iodide has been measured in the temperature range 95-225 K.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-91-4
Molecular Formula C25H25IN2
Molecular Weight (g/mol) 480.39
MDL Number MFCD00011975
InChI Key QWYZFXLSWMXLDM-UHFFFAOYSA-M
Synonym carbocyanine, pinacyanole, pinacyanol iodide, sensitol red, pinacyanol, eastman kodak 622, quinaldine blue, dsstox_cid_26576, dsstox_rid_81735, dsstox_gsid_46576
PubChem CID 5709754
ChEBI CHEBI:52218
SMILES [I-].CCN1C(=CC=CC2=CC=C3C=CC=CC3=[N+]2CC)C=CC2=CC=CC=C12

Specifications

Melting Point >250°C
Quantity 250 mg
UN Number UN2811
IUPAC Name 1-ethyl-2-[3-(1-ethyl-1,2-dihydroquinolin-2-ylidene)prop-1-en-1-yl]quinolin-1-ium iodide
Formula Weight 480.38
Percent Purity 96%
Chemical Name or Material 1,1'-Diethyl-2,2'-carbocyanine iodide
Hazard Category H301+H311-H315-H319-H330
Hazard Statement GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Precautionary Statement P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 210-099-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.