Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,1-dimethyl-1-propanol-d6 | 75295-95-3 | 94.19 g/mol | C5H6D6O | 5 MG
SDP

Supplier:  Medchemexpress LLC HYW7007425MG

Encompass_Preferred

1,1-Dimethyl-1-propanol-d6 is a deuterium-labeled analogue of 1,1-dimethyl-1-propanol used as an isotopic standard and synthetic building block in chemical and analytical research. It is supplied in small milligram quantities for laboratory use and analytical calibration. CAS number 75295-95-3; molecular formula C5H6D6O; molecular weight 94.19 g/mol.

  • Deuterium-labeled (d6) tertiary alcohol for isotope labeling and standards.
  • Suitable for NMR and mass spectrometry calibration and quantitation.
  • Supplied in milligram-scale quantities for research use.
  • Molecular formula C5H6D6O; molecular weight 94.19 g/mol.
  • Purity and storage details provided on the certificate of analysis.

Catalog No. 50-002-84700


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.