Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethylbiguanide Hydrochloride, MP Biomedicals™

Catalog No. MP215780525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215780525 25 g
ICN15780580 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP215780525 Supplier MP Biomedicals, Inc Supplier No. 0215780525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,1-Dimethylbiguanide Hydrochloride, White Powder

  • It has various medical uses such as in polycystic ovary syndrome, non-alcoholic fatty liver disease and diabetes mellitus type 2.
  • It may also reduce weight gain in people taking atypical antipsychotics. 1,1-Dimethylbiguanide hydrochloride is suitable biguanide agent used in a study to investigate its role in enhancing the secretions of plasma active glucagon-like peptide-1 (GLP-1) levels.
  • 1,1-Dimethylbiguanide, Hydrochloride (Metformin) is a biguanide antihyperglycemic agent. 1,1-Dimethylbiguanide hydrochloride (metformin) is a strong base.
  • It forms well defined salts and possessing excellent coordination capacity with transition metals, forming highly colored bidentate chelate complexes.
  • Metformin (MET) exists in various forms: diprotonated (H2MET)2+ in strong acidic solution, monoprotonated (HMET)+ in weak acid, MET in neutral and deprotonated (MET)- in strong alkali solution. It is an oral antidiabetic drug.
  • Pharmacokinetics of metformin has been studied. Determination of metformin in human plasma has been described by simple HPLC-UV method.[5] Metformin is reported to increase plasma active glucagon-like peptide-1 (GLP-1) in humans
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1115-70-4
Molecular Formula C4H12ClN5
Molecular Weight (g/mol) 165.63
MDL Number MFCD00012582
InChI Key OETHQSJEHLVLGH-UHFFFAOYSA-N
IUPAC Name hydrogen 1-[(diaminomethylidene)amino]-N,N-dimethylmethanimidamide chloride
SMILES [H+].[Cl-].CN(C)C(=N)N=C(N)N

Specifications

Melting Point 214°C to 224°C
Color White
Quantity 25 g
Synonym N,N-Dimethylimidodicarbonimidic diamine, 1,1-Dimethylbiguanide hydrochloride, DMGG, LA-6023
Solubility Information Soluble in water (100mg/mL - clear, colorless solution), 95% ethanol; insoluble in ether, acetone or chloroform.
Physical Form Powder
RTECSNumber DU1800000
storageNote1 Store at Room Temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.