Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,1-Dimethylethyl 4-(4-amino-3-pyridinyl)-1-piperazinecarboxylate | 1023594-50-4 | 99.9% | 278.36 | 100 MG
SDP

Catalog No. 5000430865
Encompass_Preferred

1,1-Dimethylethyl 4-(4-amino-3-pyridinyl)-1-piperazinecarboxylate is an impurity of Palbociclib. Palbociclib is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. This product is for research use only and not sold to patients.

  • Appearance: Solid
  • Color: Light yellow to brown
  • Target: CDK
  • Pathway: Cell Cycle/DNA Damage

Catalog No. 50-004-30865 Supplier Medchemexpress LLC Supplier No. HYI0996100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,1-Dimethylethyl 4-(4-amino-3-pyridinyl)-1-piperazinecarboxylate is an impurity of Palbociclib. Palbociclib is an orally active selective CDK4 and CDK6 inhibitor with IC50 values of 11 and 16 nM, respectively. This product is for research use only and not sold to patients.

  • Appearance: Solid
  • Color: Light yellow to brown
  • Target: CDK
  • Pathway: Cell Cycle/DNA Damage

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.