Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,1-Dimethylurea-d6 | 1219802-32-0 | 99.6% | 94.15 | C3H2D6N2O | 5 MG
SDP

Catalog No. 5000208756
Encompass_Preferred

1,1-Dimethylurea-d6 is the deuterium-labeled isotopologue of 1,1-dimethylurea used as a tracer or internal standard in analytical chemistry and mass spectrometry. It is supplied as a white to off-white solid in small research-scale quantities.

  • Purity 99.6% (HPLC)
  • Molecular weight 94.15 g/mol
  • Chemical formula C3H2D6N2O
  • Appearance white to off-white solid
  • Available in 1 mg, 5 mg, and 10 mg pack sizes
  • Used as tracer and internal standard in analytical methods

Catalog No. 50-002-08756 Supplier Medchemexpress LLC Supplier No. HYW089832S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,1-Dimethylurea-d6 is the deuterium-labeled isotopologue of 1,1-dimethylurea used as a tracer or internal standard in analytical chemistry and mass spectrometry. It is supplied as a white to off-white solid in small research-scale quantities.

  • Purity 99.6% (HPLC)
  • Molecular weight 94.15 g/mol
  • Chemical formula C3H2D6N2O
  • Appearance white to off-white solid
  • Available in 1 mg, 5 mg, and 10 mg pack sizes
  • Used as tracer and internal standard in analytical methods

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.