Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dimethylurea 98.0+%, TCI America™
SDP

Catalog No. D080925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D080925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D080925G Supplier TCI America Supplier No. D080925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 598-94-7
Melting Point 182°C
Color White
Molecular Formula C3H8N2O
Quantity 25 g
MDL Number MFCD00007959
Synonym n,n-dimethylurea, urea, n,n-dimethyl, asym-dimethylurea, urea, dimethyl, urea, 1,1-dimethyl, n,n-dimethylharnstoff german, n,n-dimethylharnstoff, dimethyl urea, 1.1-dimethylurea, 1,1-dimethyl-urea
InChI Key YBBLOADPFWKNGS-UHFFFAOYSA-N
SMILES CN(C)C(=O)N
IUPAC Name 1,1-dimethylurea
Molecular Weight (g/mol) 88.11
PubChem CID 11737
Formula Weight 88.11
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Dimethylurea
Show More Show Less
EINECSNumber (2)-3366
RTECSNumber YS9867985
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.