Learn More
1,1'-Dioctadecyl-3,3,3',3'-Tetramethylindocarbocyanine Perchlorate, Sigma-Aldrich
Shop All MilliporeSigma Supelco ProductsDescription
- Useful for optical recordings of membrane voltage and membrane-fluidity studies.
- Provides persistent staining for retrograde neuronal labeling.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | Stain |
| Molecular Weight (g/mol) | 933.87 g/mol |
| Purity | 97% |
| Label Type | Other Labels or Dyes |
| Color | Dark Purple |
| CAS | 41085-99-8 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.