Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Dioxobenzo[b]thiophen-2-ylmethyl chloroformate, tech. 90%, Thermo Scientific™

Catalog No. AAL1955903
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1955903 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1955903 Supplier Thermo Scientific Chemicals Supplier No. L1955903

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1,1-Dioxobenzo[b]thiophen-2-ylmethyl chloroformate is used for protection of amino acids as their benzothiophenesulfone-2-methoxycarbonyl (Bsmoc) derivatives, which represent a significant improvement in solid-phase peptide synthesis over the corresponding Fmoc derivatives.

Solubility
Reacts with water.

Notes
Store under inert gas. It is sensitive to moisture. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 135204-19-2
Molecular Formula C10H7ClO4S
Quantity 1g
MDL Number MFCD01090984
UN Number UN3261
Beilstein 8258946
Synonym 1,1-dioxobenzo b thiophen-2-ylmethyl chloroformate, 1,1-dioxidobenzo b thiophen-2-yl methyl carbonochloridate, bsmoc-cl, acmc-20ap7q, 1-dioxobenzo b thiophen-2-ylmethyl chloroformate, benzo b thiophenesulfone-2-methoxycarbonyl chloride, 2-chlorocarbonyloxymethyl benzo b thiophene 1,1-dioxide, 1,1-dioxo-1-benzothiophen-2-yl methyl carbonochloridate, 1,1-dioxo-1??-benzothiophen-2-yl methyl carbonochloridate, 1,1-dioxo-1??-benzothiophen-2-yl methyl chloroformate
InChI Key ZYXGPSYADVTJGF-UHFFFAOYSA-N
SMILES ClC(=O)OCC1=CC2=CC=CC=C2S1(=O)=O
IUPAC Name (1,1-dioxo-1λ⁶-benzothiophen-2-yl)methyl carbonochloridate
Molecular Weight (g/mol) 258.67
PubChem CID 4565178
Formula Weight 258.68
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,1-Dioxobenzo[b]thiophen-2-ylmethyl chloroformate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Recommended Storage 2° to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.