Learn More
Medchemexpress LLC 1,1-Dioxothiolan-d8 | 51219-88-6 | 99.9% | 128.22 | 5 MG
SDP
Catalog No. 5000404121
Encompass_Preferred
1,1-Dioxothiolan-d8 is the deuterium labeled 1,1-Dioxothiolan. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotopes for drug development process
  • Potential to affect pharmacokinetic and metabolic profiles
Catalog No. 50-004-04121 Supplier Medchemexpress LLC Supplier No. HY34444S5MG
1,1-Dioxothiolan-d8 is the deuterium labeled 1,1-Dioxothiolan. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Stable heavy isotopes for drug development process
  • Potential to affect pharmacokinetic and metabolic profiles