Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diphenyl-4-piperidino-1-butanol Hydrochloride 99.0+%, TCI America™
SDP

Catalog No. D206225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D206225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D206225G Supplier TCI America Supplier No. D206225G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3254-89-5
Molecular Formula C21H28ClNO
Molecular Weight (g/mol) 345.911
MDL Number MFCD00151479
InChI Key AVZIYZHXZAYGJS-UHFFFAOYSA-N
Synonym Diphenidol Hydrochloride, 1-(4-Hydroxy-4,4-diphenylbutyl)piperidine Hydrochloride
PubChem CID 66266
IUPAC Name 1,1-diphenyl-4-piperidin-1-ylbutan-1-ol;hydrochloride
SMILES C1CCN(CC1)CCCC(C2=CC=CC=C2)(C3=CC=CC=C3)O.Cl

Specifications

Melting Point 214°C
Color White
Quantity 25 g
Formula Weight 345.91
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,1-Diphenyl-4-piperidino-1-butanol Hydrochloride
EINECSNumber (9)-0255
RTECSNumber TM4970500
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.