Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Diphenylhydrazine 97.0+%, TCI America™
SDP

Catalog No. D11615ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D11615ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D11615ML Supplier TCI America Supplier No. D11615ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 530-50-7
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD00044505
InChI Key YHYKLKNNBYLTQY-UHFFFAOYSA-N
PubChem CID 10739
IUPAC Name 1,1-diphenylhydrazine
SMILES C1=CC=C(C=C1)N(C2=CC=CC=C2)N

Specifications

Melting Point 35°C
Boiling Point 220°C
Quantity 5 mL
Formula Weight 184.24
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 1,1-Diphenylhydrazine
RTECSNumber MV3450000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.