Learn More
Medchemexpress LLC (1,1'-Dipyrenyl) dimethyl ether | 74833-81-1 | 98.0% | 446.54 | 1 MG
SDP
Catalog No. 5000362519
Encompass_Preferred
(1,1'-Dipyrenyl)dimethyl ether is a fluorescent dye that exhibits intramolecular excimer fluorescence. It is also soluble in synthetic phospholipid membranes and competes with fluorescence from the locally excited pyrene chromophore.
  • Exhibits intramolecular excimer fluorescence
  • Soluble in synthetic phospholipid membranes
  • Functions as a fluorescent dye
Catalog No. 50-003-62519 Supplier Medchemexpress LLC Supplier No. HY1471701MG
(1,1'-Dipyrenyl)dimethyl ether is a fluorescent dye that exhibits intramolecular excimer fluorescence. It is also soluble in synthetic phospholipid membranes and competes with fluorescence from the locally excited pyrene chromophore.
  • Exhibits intramolecular excimer fluorescence
  • Soluble in synthetic phospholipid membranes
  • Functions as a fluorescent dye