Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (1,1'-Dipyrenyl) dimethyl ether | 74833-81-1 | 98.0% | 446.54 | 5 MG
SDP

Catalog No. 5000360225
Encompass_Preferred

(1,1'-Dipyrenyl)dimethyl ether exhibits intramolecular excimer fluorescence in competition with fluorescence from the locally excited pyrene chromophore. It is soluble in synthetic phospholipid membranes.

  • Exhibits intramolecular excimer fluorescence.
  • Fluorescence competes with locally excited pyrene chromophore.
  • Soluble in synthetic phospholipid membranes.

Catalog No. 50-003-60225 Supplier Medchemexpress LLC Supplier No. HY1471705MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(1,1'-Dipyrenyl)dimethyl ether exhibits intramolecular excimer fluorescence in competition with fluorescence from the locally excited pyrene chromophore. It is soluble in synthetic phospholipid membranes.

  • Exhibits intramolecular excimer fluorescence.
  • Fluorescence competes with locally excited pyrene chromophore.
  • Soluble in synthetic phospholipid membranes.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.