Learn More
Medchemexpress LLC 1-(1-ethylpiperidin-4-yl)sulfonyl-3-(1,2,3,5,6,7-hexahydro-s-indacen-4-yl)urea | 2260969-36-4 | MFCD34579587 | 99.5% | C20H29N3O3S | 10MG
SDP
Catalog No. 5000261706
Encompass_Preferred
Selnoflast is an orally active, potent, selective, and reversible small-molecule inhibitor of the NLRP3 inflammasome for research use. It is supplied as a white to off-white solid with high reported purity, and the manufacturer provides solubility and storage guidance for preparation and stability in in vitro and preclinical workflows.
  • Potent and selective NLRP3 inflammasome inhibition.
  • Reversible small-molecule mechanism enabling temporal studies.
  • High reported purity for consistent experimental results.
  • Solid form suitable for standard laboratory storage and handling.
  • Documented solubility in DMSO and alkaline media with preparation notes.
  • Specified storage recommendations for powder and solution stability.
  • Multiple pack sizes available to support different study scales.
Catalog No. 50-002-61706 Supplier Medchemexpress LLC Supplier No. HY13283110MG
Selnoflast is an orally active, potent, selective, and reversible small-molecule inhibitor of the NLRP3 inflammasome for research use. It is supplied as a white to off-white solid with high reported purity, and the manufacturer provides solubility and storage guidance for preparation and stability in in vitro and preclinical workflows.
  • Potent and selective NLRP3 inflammasome inhibition.
  • Reversible small-molecule mechanism enabling temporal studies.
  • High reported purity for consistent experimental results.
  • Solid form suitable for standard laboratory storage and handling.
  • Documented solubility in DMSO and alkaline media with preparation notes.
  • Specified storage recommendations for powder and solution stability.
  • Multiple pack sizes available to support different study scales.