Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Methylene-di-2-naphthol, 98%, Thermo Scientific™

Catalog No. AC330600250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC330600250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC330600250 Supplier Thermo Scientific Chemicals Supplier No. 330600250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1096-84-0
Molecular Formula C21H16O2
Molecular Weight (g/mol) 300.357
InChI Key ZPANWZBSGMDWON-UHFFFAOYSA-N
Synonym 1,1'-methylenedi-2-naphthol, squoxin, 1,1'-methylenebis naphthalen-2-ol, 2-naphthalenol, 1,1'-methylenebis, 1,1'-methylenebis 2-naphthol, 1,1-methylenedi-2-naphthol, unii-wse97k4l90, 2-naphthol, 1,1'-methylenedi, bis 2-hydroxy-1-naphthyl methane, 1-2-hydroxy-1-naphthyl methyl naphthalen-2-ol
PubChem CID 14187
IUPAC Name 1-[(2-hydroxynaphthalen-1-yl)methyl]naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=CC(=C2CC3=C(C=CC4=CC=CC=C43)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Pink to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Quantity 25 g
Beilstein 06, IV, 7025
Formula Weight 300.36
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 1, 1-Methylene-di-2-naphthol, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.