Learn More
Medchemexpress LLC 1-(1-methylindol-5-yl)-3-(3-methyl-1,2-thiazol-5-yl)urea | 152239-46-8 | MFCD00923644 | 99.9% | 286.35 g/mol | C14H14N4OS | 50 MG
SDP
Catalog No. 5000245925
Encompass_Preferred
SB 204741 is a selective, high-affinity antagonist of the serotonin 5-HT2B receptor supplied for research use. It is used in pharmacology studies to probe 5-HT2B-mediated signaling and receptor function.
  • Selective 5-HT2B receptor antagonist for receptor pharmacology studies.
  • High affinity for 5-HT2B (pKi ≈ 7.1), enabling sensitive receptor profiling.
  • High purity (~99.9%) and defined molecular properties (C14H14N4OS, 286.35 g/mol).
  • Available in small solid quantities suitable for in vitro and in vivo research.
Catalog No. 50-002-45925 Supplier Medchemexpress LLC Supplier No. HY10315350MG
SB 204741 is a selective, high-affinity antagonist of the serotonin 5-HT2B receptor supplied for research use. It is used in pharmacology studies to probe 5-HT2B-mediated signaling and receptor function.
  • Selective 5-HT2B receptor antagonist for receptor pharmacology studies.
  • High affinity for 5-HT2B (pKi ≈ 7.1), enabling sensitive receptor profiling.
  • High purity (~99.9%) and defined molecular properties (C14H14N4OS, 286.35 g/mol).
  • Available in small solid quantities suitable for in vitro and in vivo research.