Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Naphthyl)ethanol 98.0+%, TCI America™
SDP

Catalog No. N03951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N03951G 1 g
N03955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N03951G Supplier TCI America Supplier No. N03951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1517-72-2
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.227
MDL Number MFCD00016806
InChI Key CDRQOYRPWJULJN-UHFFFAOYSA-N
Synonym 1-(1-Hydroxyethyl)naphthalene, alpha-Methyl-1-naphthalenemethanol
PubChem CID 98030
IUPAC Name 1-naphthalen-1-ylethanol
SMILES CC(C1=CC=CC2=CC=CC=C21)O

Specifications

Melting Point 64°C
Color White-Yellow
Boiling Point 133°C
Quantity 1 g
Formula Weight 172.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(1-Naphthyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.