Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+)-1-(1-Naphthyl)ethylamine | 3886-70-2 | 99.99% | C12H13N | 25 G
SDP

Catalog No. 5000358459
Encompass_Preferred

(+)-1-(1-Naphthyl)ethylamine is a chiral modifier utilized to introduce enantioselectivity in catalytic hydrogenation reactions. This compound can undergo H-D exchange with D2 in solution to form N-D bonds, indicating its potential to impart enantioselectivity to Pt hydrogenation catalysts by completing Pt surface adsorption and protonation through amine N atoms.

  • Appears as a liquid
  • Density: 1.064±0.06 g/cm3
  • Color: Colorless to light yellow

Catalog No. 50-003-58459 Supplier Medchemexpress LLC Supplier No. 25GHY75070
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(+)-1-(1-Naphthyl)ethylamine is a chiral modifier utilized to introduce enantioselectivity in catalytic hydrogenation reactions. This compound can undergo H-D exchange with D2 in solution to form N-D bonds, indicating its potential to impart enantioselectivity to Pt hydrogenation catalysts by completing Pt surface adsorption and protonation through amine N atoms.

  • Appears as a liquid
  • Density: 1.064±0.06 g/cm3
  • Color: Colorless to light yellow

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.