Learn More
Medchemexpress LLC (+)-1-(1-Naphthyl)ethylamine | 3886-70-2 | 99.99% | C12H13N | 50 G
SDP
Catalog No. 5000434228
Encompass_Preferred
(+)-1-(1-Naphthyl)ethylamine ((+)-1-(1-NEA)) is a chiral modifier utilized to introduce enantioselectivity in catalytic hydrogenation reactions. This compound, 1-NEA, can undergo H-D exchange with D2 in solution to form N-D bonds. This indicates that 1-NEA has the capability to complete Pt surface adsorption and protonation through amine N atoms, thereby potentially imparting enantioselectivity to Pt hydrogenation catalysts.
  • Utilized to introduce enantioselectivity in catalytic hydrogenation reactions.
  • Can undergo H-D exchange with D2 in solution to form N-D bonds.
  • Capable of completing Pt surface adsorption and protonation through amine N atoms.
  • Potentially imparts enantioselectivity to Pt hydrogenation catalysts.
Catalog No. 50-004-34228 Supplier Medchemexpress LLC Supplier No. HY7507050G
(+)-1-(1-Naphthyl)ethylamine ((+)-1-(1-NEA)) is a chiral modifier utilized to introduce enantioselectivity in catalytic hydrogenation reactions. This compound, 1-NEA, can undergo H-D exchange with D2 in solution to form N-D bonds. This indicates that 1-NEA has the capability to complete Pt surface adsorption and protonation through amine N atoms, thereby potentially imparting enantioselectivity to Pt hydrogenation catalysts.
  • Utilized to introduce enantioselectivity in catalytic hydrogenation reactions.
  • Can undergo H-D exchange with D2 in solution to form N-D bonds.
  • Capable of completing Pt surface adsorption and protonation through amine N atoms.
  • Potentially imparts enantioselectivity to Pt hydrogenation catalysts.