Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1-Oxybis[3-(2-methoxyphenoxy)-2-propanol] Pharmaceutical Impurity Standard, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002492 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2492 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2492 Supplier MilliporeSigma Supelco Supplier No. 7921350MG
Add to Cart
Add to Cart
Featured Industries: Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 1797132-23-0
Linear Formula C20H26O7
MDL Number MFCD01631116
Synonym Guaifenesin impurity C (PhEur)
Type Pharmaceutical Impurity Standard
Quantity 50 mg
Formula Weight 378.42
Percent Purity ≥98.0% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.