Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(1-Phenylethyl)piperazine hydrochloride | 436099-96-6 | 95.0% | 226.75 | 1 G
SDP

Catalog No. 5000405207
Encompass_Preferred

1-(1-Phenylethyl)piperazine hydrochloride is a drug intermediate used in the synthesis of various active compounds.

  • Target: Drug Intermediate
  • Pathway: Others
  • Storage: Store at room temperature 3 years; in solvent -80°C 2 years, -20°C 1 year.

Catalog No. 50-004-05207 Supplier Medchemexpress LLC Supplier No. HY442651G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(1-Phenylethyl)piperazine hydrochloride is a drug intermediate used in the synthesis of various active compounds.

  • Target: Drug Intermediate
  • Pathway: Others
  • Storage: Store at room temperature 3 years; in solvent -80°C 2 years, -20°C 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.