Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(1-Phenylethyl)piperazine hydrochloride | 436099-96-6 | 95.0% | 226.75 | 100 MG
SDP

Catalog No. 5000403864
Encompass_Preferred

1-(1-Phenylethyl)piperazine hydrochloride is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Drug intermediate for the synthesis of various active compounds.
  • For research use only.
  • Molecular weight: 226.75
  • Formula: C12H19ClN2
  • Appearance: Solid, white to off-white.
  • Store at room temperature for 3 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.

Catalog No. 50-004-03864 Supplier Medchemexpress LLC Supplier No. HY44265100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(1-Phenylethyl)piperazine hydrochloride is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Drug intermediate for the synthesis of various active compounds.
  • For research use only.
  • Molecular weight: 226.75
  • Formula: C12H19ClN2
  • Appearance: Solid, white to off-white.
  • Store at room temperature for 3 years.
  • Store in solvent at -80°C for 2 years or -20°C for 1 year.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.