Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(1-Phenylethyl)piperazine hydrochloride | 436099-96-6 | 95.0% | 226.75 | 250 MG
SDP

Catalog No. 5000402742
Encompass_Preferred

1-(1-Phenylethyl)piperazine hydrochloride is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Drug intermediate for the synthesis of various active compounds.
  • For research use only.
  • Appears as a white to off-white solid.
  • Store at room temperature for up to 3 years.
  • Soluble in DMSO (250 mg/mL).
  • Protocols available for in vivo solubility in various solvent systems.

Catalog No. 50-004-02742 Supplier Medchemexpress LLC Supplier No. 250MGHY44265
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-(1-Phenylethyl)piperazine hydrochloride is a drug intermediate for the synthesis of various active compounds. It is for research use only and not sold to patients.

  • Drug intermediate for the synthesis of various active compounds.
  • For research use only.
  • Appears as a white to off-white solid.
  • Store at room temperature for up to 3 years.
  • Soluble in DMSO (250 mg/mL).
  • Protocols available for in vivo solubility in various solvent systems.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.