Learn More
Medchemexpress LLC 1-(1-Phenylethyl)piperazine hydrochloride | 436099-96-6 | 95.0% | 226.75 | 5 G
SDP
Catalog No. 5000403830
Encompass_Preferred
1-(1-Phenylethyl)piperazine hydrochloride is a drug intermediate used for the synthesis of various active compounds. This product is provided as a white to off-white solid.
  • High purity solid form
  • Stable for 3 years at room temperature
  • Soluble in DMSO for in vitro applications
  • Multiple in vivo solubility protocols available
Catalog No. 50-004-03830 Supplier Medchemexpress LLC Supplier No. HY442655G
1-(1-Phenylethyl)piperazine hydrochloride is a drug intermediate used for the synthesis of various active compounds. This product is provided as a white to off-white solid.
  • High purity solid form
  • Stable for 3 years at room temperature
  • Soluble in DMSO for in vitro applications
  • Multiple in vivo solubility protocols available