Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(1-Pyrenyl)ethanol, 98%, Thermo Scientific™

Catalog No. AC449290010
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449290010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC449290010 Supplier Thermo Scientific Chemicals Supplier No. 449290010
Add to Cart

Chemical Identifiers

CAS 65954-42-9
Molecular Formula C18H14O
Molecular Weight (g/mol) 246.31
InChI Key LFCWFVPSUMBNQM-UHFFFAOYSA-N
Synonym 1-1-pyrenyl ethanol, 1-hydroxyethylpyrene, alpha-methyl-1-pyrenemethanol, 1-1-hydroxyethyl pyrene, ccris 7293, 1-pyrenemethanol, alpha-methyl, 1-pyren-1-yl ethanol, 1-pyrenylethan-1-ol, 4-06-00-05012 beilstein handbook reference
PubChem CID 119118
IUPAC Name 1-pyren-1-ylethanol
SMILES CC(C1=C2C=CC3=CC=CC4=C3C2=C(C=C4)C=C1)O

Specifications

Melting Point 111.0°C to 113.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 246.31
Percent Purity 98%
Chemical Name or Material 1-(1-Pyrenyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.