Learn More
Medchemexpress LLC HY-100740C 5mg Medchemexpress, (1α,1'S,4β)-Lanabecestat CAS:1384082-96-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100740C5MG
Encompass_Preferred
Medchemexpress, HY-100740C 5mg (1α,1'S,4β)-Lanabecestat CAS:1384082-96-5 (1α,1'S,4β)-Lanabecestat ((1α,1'S,4β)-AZD3293) is a Beta site APP Cleaving Enzymel (BACE1) inhibitor extracted from patent WO2012087237A1, compound 20a, has IC50s of 2.2 nM (TR-FRET assay) and 0.28 nM (sAPPp release assay), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7777