Learn More
ANALYTICAL SCIENTIFIC INSTRUMENTS US INC ASI 610-PO10-06 Post-Column Splitter, Adjustable Flow Split Ratio 1:1 to 20:1, Maximum Pressure 350 bar, 4.8 in x 4 in x 5.1 in
SDP
Catalog No. NC1916809
Encompass
The ASI 610-PO10-06 is an analytical post-column adjustable flow splitter designed for High-Performance Liquid Chromatography (HPLC). It excels in providing precise flow splitting with a range of 1:1 to 20:1 and is stable under varying solvent conditions. This product's compact design and hand adjustment knob enhance user convenience, setting it apart in optimizing flow rates efficiently.
Catalog No. NC1916809 Supplier ANALYTICAL SCIENTIFIC INSTRUMENTS US INC Supplier No. 610PO1006
The ASI 610-PO10-06 is an analytical post-column adjustable flow splitter designed for High-Performance Liquid Chromatography (HPLC). It excels in providing precise flow splitting with a range of 1:1 to 20:1 and is stable under varying solvent conditions. This product's compact design and hand adjustment knob enhance user convenience, setting it apart in optimizing flow rates efficiently.