Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,10-bis(pyridinium)decane | 6266-40-6 | MFCD00031922 | 99.7% | 458.27 g/mol | C20H30Br2N2 | 5 MG
SDP

Catalog No. 5000280940
Encompass_Preferred

1,10-Bis(pyridinium)decane is a biscationic pyridinium salt used as a biochemical research reagent. It acts as an acetylcholinesterase (AChE) inhibitor with a reported IC50 of 25.8 nM and is supplied at high purity for in vitro pharmacology and enzymology studies. Available in milligram-scale quantities for laboratory use.

  • Bicationic pyridinium salt suitable for cholinesterase research
  • Reported AChE IC50 of 25.8 nM
  • High purity: 99.66%
  • Molecular weight: 458.27 g/mol
  • Chemical formula: C20H30Br2N2
  • Supplied in milligram-scale pack sizes for laboratory experiments
  • For research use only

Catalog No. 50-002-80940 Supplier Medchemexpress LLC Supplier No. HYW1650855MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,10-Bis(pyridinium)decane is a biscationic pyridinium salt used as a biochemical research reagent. It acts as an acetylcholinesterase (AChE) inhibitor with a reported IC50 of 25.8 nM and is supplied at high purity for in vitro pharmacology and enzymology studies. Available in milligram-scale quantities for laboratory use.

  • Bicationic pyridinium salt suitable for cholinesterase research
  • Reported AChE IC50 of 25.8 nM
  • High purity: 99.66%
  • Molecular weight: 458.27 g/mol
  • Chemical formula: C20H30Br2N2
  • Supplied in milligram-scale pack sizes for laboratory experiments
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.