Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Decanedicarbonitrile, 98%, Thermo Scientific™

Catalog No. AAL0345106
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0345106 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0345106 Supplier Thermo Scientific Chemicals Supplier No. L0345106

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The Dodecanedinitrile could react with C-tert-butoxy-tetra-N-methyl-methanediamine to obtain the 2-(Dimethylaminomethylen)dodecansaeuredinitril.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 4543-66-2
Density 0.898
Boiling Point 195°C (8mmHg)
Flash Point 202°C (395°F)
Molecular Formula C12H20N2
Refractive Index 1.451
Quantity 5g
MDL Number MFCD00013818
Beilstein 1757752
Synonym 1,10-dicyanodecane, decamethylenedinitrile, dodecanodinitrile, 1,10-decanedicarbonitrile, dodecandinitril, 1,12-dodecanedinitrile, 1,10-decamethylenedinitrile, decamethylene dicyanide, decane-1,10-dicarboxylic dinitrile, dodecamethylene dinitrile
Solubility Information Insoluble in water.
InChI Key AVQYXBDAZWIFTO-UHFFFAOYSA-N
SMILES C(CCCCCC#N)CCCCC#N
IUPAC Name dodecanedinitrile
Molecular Weight (g/mol) 192.306
PubChem CID 78301
Formula Weight 192.3
Percent Purity 98%
Chemical Name or Material 1,10-Decanedicarbonitrile
Show More Show Less
EINECSNumber 224-899-6
RTECSNumber JR2350000
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.