Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,10-Decanediol dimethacrylate

Catalog No. 8087255G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
80-872-55G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 80-872-55G Supplier Sigma Aldrich Supplier No. 8087255G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,10-decamethylene glycol dimethacrylate is a monomer that can be used to make materials for use in dental applications. It could be used to prepare camphorquinone/amine based adhesive.

Specifications

CAS 6701-13-9
Density 0.968 g/mL (at 25°C)
Molecular Formula C18H30O4
Refractive Index n/D 1.462
Linear Formula C18H30O4
Quantity 5 g
MDL Number MFCD00080452
Synonym 2-Propenoic acid; 2-methyl-1,1 -(1,10-decanediyl) ester; Decamethylene glycol dimethacrylate
Molecular Weight (g/mol) 310.43
Percent Purity 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.