Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,10-Diacetoxydecane 97.0+%, TCI America™
SDP

Catalog No. D278325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D278325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D278325G Supplier TCI America Supplier No. D278325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 26118-61-6
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
MDL Number MFCD03093628
InChI Key NMOLZWLFORPIME-UHFFFAOYSA-N
Synonym Decamethylene Diacetate, 1,10-Decanediol Diacetate
PubChem CID 117736
IUPAC Name 10-acetyloxydecyl acetate
SMILES CC(=O)OCCCCCCCCCCOC(=O)C

Specifications

Color White
Quantity 25 g
Formula Weight 258.36
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1,10-Diacetoxydecane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.