Learn More
Medchemexpress LLC 1,10-Dibromodecane-d20 | 150017-88-2 | 98.1% | 320.20 | 10 MG
SDP
Catalog No. 5000366918
Encompass_Preferred
1,10-Dibromodecane-d20 is the deuterium labeled 1,10-Dibromodecan. It is for research use only and not sold to patients. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-66918 Supplier Medchemexpress LLC Supplier No. HYW009240S10MG
1,10-Dibromodecane-d20 is the deuterium labeled 1,10-Dibromodecan. It is for research use only and not sold to patients. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, has potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.