Learn More
Medchemexpress LLC 1,10-Dibromodecane-d20 | 150017-88-2 | 98.1% | 320.20 | 5 MG
SDP
Catalog No. 5000366846
Encompass_Preferred
This product is a deuterium labeled 1,10-Dibromodecane, intended for research use only. It incorporates stable heavy isotopes of hydrogen, carbon, and other elements. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Appearance: liquid
  • Color: colorless to light yellow
Catalog No. 50-003-66846 Supplier Medchemexpress LLC Supplier No. HYW009240S5MG
This product is a deuterium labeled 1,10-Dibromodecane, intended for research use only. It incorporates stable heavy isotopes of hydrogen, carbon, and other elements. Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Appearance: liquid
  • Color: colorless to light yellow