Learn More
Medchemexpress LLC 1 10-Diiododecane 100g
SDP
Catalog No. 5000206937
Encompass_Preferred
1 10-Diiododecane (Decamethylene diiodide) serves as a crosslinking reagent for tetrabutylammonium polygalacturonic acid and acts as an alkylating agent in the synthesis of both symmetrical and unsymmetrical bis-cryptophanes in addition to being utilized for the production of decane at a temperature of 20 C
Catalog No. 50-002-06937 Supplier Medchemexpress LLC Supplier No. HYW105731100G
1 10-Diiododecane (Decamethylene diiodide) serves as a crosslinking reagent for tetrabutylammonium polygalacturonic acid and acts as an alkylating agent in the synthesis of both symmetrical and unsymmetrical bis-cryptophanes in addition to being utilized for the production of decane at a temperature of 20 C