Learn More
Medchemexpress LLC 1 10-Diiododecane 50g
SDP
Catalog No. 5000205025
Encompass_Preferred
1 10-Diiododecane (Decamethylene diiodide) serves as a crosslinking reagent for tetrabutylammonium polygalacturonic acid and acts as an alkylating agent in the synthesis of both symmetrical and unsymmetrical bis-cryptophanes in addition to being utilized for the production of decane at a temperature of 20 C
Catalog No. 50-002-05025 Supplier Medchemexpress LLC Supplier No. HYW10573150G
1 10-Diiododecane (Decamethylene diiodide) serves as a crosslinking reagent for tetrabutylammonium polygalacturonic acid and acts as an alkylating agent in the synthesis of both symmetrical and unsymmetrical bis-cryptophanes in addition to being utilized for the production of decane at a temperature of 20 C