Learn More
Alkali Scientific 1, 10 Phenanthroline, 10 G
SDP
Catalog No. 5000661829
Encompass_Preferred
1,10-Phenanthroline is a high-purity chelating agent used in analytical chemistry for the determination of metal ions, particularly in complexometric titrations. This reagent forms stable complexes with divalent metal ions, making it valuable in metal ion analysis and the study of coordination chemistry. The compound is commonly used in biochemistry and molecular biology research to chelate metal ions in protein assays, enzyme studies, and DNA analysis. 1,10-Phenanthroline is also essential in environmental chemistry for monitoring metal contamination. Provided in a 10g quantity, it is ideal for high-precision analytical work in research labs requiring accurate and reliable metal ion analysis.
Catalog No. 50-006-61829 Supplier Alkali Scientific Supplier No. XXX001CP
1,10-Phenanthroline is a high-purity chelating agent used in analytical chemistry for the determination of metal ions, particularly in complexometric titrations. This reagent forms stable complexes with divalent metal ions, making it valuable in metal ion analysis and the study of coordination chemistry. The compound is commonly used in biochemistry and molecular biology research to chelate metal ions in protein assays, enzyme studies, and DNA analysis. 1,10-Phenanthroline is also essential in environmental chemistry for monitoring metal contamination. Provided in a 10g quantity, it is ideal for high-precision analytical work in research labs requiring accurate and reliable metal ion analysis.