Learn More
Research Products International Corp 1 10-Phenanthroline 10 G
SDP
Catalog No. 501978247
Encompass_Preferred
Forms a complex with ferrous ions. It can be used as an indicator in oxidation-reduction systems in fitrating ferrous salts. Material passes test for redox indicator and passes test for iron determination.
Catalog No. 50-197-8247 Supplier Research Products International Corp Supplier No. P3020010.0
Forms a complex with ferrous ions. It can be used as an indicator in oxidation-reduction systems in fitrating ferrous salts. Material passes test for redox indicator and passes test for iron determination.