Learn More
Medchemexpress LLC 1,10-Phenanthroline, 3,4,7,8-tetramethyl- | 1660-93-1 | 99.5% | 236.32 | 1 G
SDP
Catalog No. 5000405062
Encompass_Preferred
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule commonly used as a ligand or catalyst. It has diverse applications in organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices. Due to its performance in fluorescent probes, biosensors, and photocatalytic reactions, it is widely used in chemistry and life sciences research. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Commonly used as a ligand or catalyst
  • Wide range of applications including organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices
  • Exhibits excellent performance in fluorescent probes, biosensors, and photocatalytic reactions
  • Widely utilized in chemistry and life sciences research
Catalog No. 50-004-05062 Supplier Medchemexpress LLC Supplier No. HY345151G
3,4,7,8-Tetramethyl-1,10-phenanthroline (TMPhen) is an organic molecule commonly used as a ligand or catalyst. It has diverse applications in organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices. Due to its performance in fluorescent probes, biosensors, and photocatalytic reactions, it is widely used in chemistry and life sciences research. It is a biochemical reagent that can be used as a biological material or organic compound for life science related research.
  • Commonly used as a ligand or catalyst
  • Wide range of applications including organometallic chemical reactions, electrochemical detection, and organic optoelectronic devices
  • Exhibits excellent performance in fluorescent probes, biosensors, and photocatalytic reactions
  • Widely utilized in chemistry and life sciences research