Learn More
Medchemexpress LLC 1,10-Phenanthroline | 66-71-7 | 180.21 | 1 G
SDP
Catalog No. 5000445469
Encompass_Preferred
o-Phenanthroline (1,10-Phenanthroline) is a metal chelator and an MMP inhibitor. It prevents the induction of chromosomal aberrations in streptozotocin-treated cells and forms a red chelate with Fe2+ that absorbs maximally at 510 nm. This product is for research use only and has not been fully validated for medical applications.
Catalog No. 50-004-45469 Supplier Medchemexpress LLC Supplier No. HYW0045441G
Add to Cart
Add to Cart
o-Phenanthroline (1,10-Phenanthroline) is a metal chelator and an MMP inhibitor. It prevents the induction of chromosomal aberrations in streptozotocin-treated cells and forms a red chelate with Fe2+ that absorbs maximally at 510 nm. This product is for research use only and has not been fully validated for medical applications.