Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,10-Phenanthroline | 66-71-7 | 180.21 | 10 G
SDP

Catalog No. 5000445397
Encompass_Preferred

1,10-Phenanthroline is a metal chelator that prevents the induction of chromosomal aberrations in streptozotocin-treated cells. It forms a red chelate with Fe2+ that absorbs maximally at 510 nm and is also identified as a matrix metalloprotease (MMP) inhibitor.

  • Metal chelator
  • Prevents induction of chromosomal aberrations in streptozotocin-treated cells
  • Forms a red chelate with Fe2+ that absorbs maximally at 510 nm
  • Matrix metalloprotease (MMP) inhibitor
  • Purity of 99.96%

Catalog No. 50-004-45397 Supplier Medchemexpress LLC Supplier No. HYW00454410G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1,10-Phenanthroline is a metal chelator that prevents the induction of chromosomal aberrations in streptozotocin-treated cells. It forms a red chelate with Fe2+ that absorbs maximally at 510 nm and is also identified as a matrix metalloprotease (MMP) inhibitor.

  • Metal chelator
  • Prevents induction of chromosomal aberrations in streptozotocin-treated cells
  • Forms a red chelate with Fe2+ that absorbs maximally at 510 nm
  • Matrix metalloprotease (MMP) inhibitor
  • Purity of 99.96%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.