Learn More
Medchemexpress LLC 1,10-Phenanthroline | 66-71-7 | 180.21 | 5 G
SDP
Catalog No. 5000445468
Encompass_Preferred
1,10-Phenanthroline is a metal chelator and MMP inhibitor. It prevents the induction of chromosomal aberrations in streptozotocin-treated cells and forms a red chelate with Fe2+ that absorbs maximally at 510 nm.
  • Metal chelator
  • Prevents induction of chromosomal aberrations in streptozotocin-treated cells
  • Forms a red chelate with Fe2+ that absorbs maximally at 510 nm
  • MMP inhibitor
Catalog No. 50-004-45468 Supplier Medchemexpress LLC Supplier No. HYW0045445G
1,10-Phenanthroline is a metal chelator and MMP inhibitor. It prevents the induction of chromosomal aberrations in streptozotocin-treated cells and forms a red chelate with Fe2+ that absorbs maximally at 510 nm.
  • Metal chelator
  • Prevents induction of chromosomal aberrations in streptozotocin-treated cells
  • Forms a red chelate with Fe2+ that absorbs maximally at 510 nm
  • MMP inhibitor