Learn More
Medchemexpress LLC 1,10-Phenanthroline | 66-71-7 | 180.21 | 500 MG
SDP
Catalog No. 5000445470
Encompass_Preferred
1,10-Phenanthroline, also known as o-Phenanthroline, is a potent metal chelator and MMP inhibitor. It has been shown to prevent the induction of chromosomal aberrations in streptozotocin-treated cells. This compound forms a characteristic red chelate with Fe2+, exhibiting maximum absorption at 510 nm.
  • Acts as a metal chelator
  • Prevents chromosomal aberrations in streptozotocin-treated cells
  • Forms a red chelate with Fe2+ with maximum absorption at 510 nm
  • Functions as an MMP inhibitor
  • Appears as a white to off-white solid
Catalog No. 50-004-45470 Supplier Medchemexpress LLC Supplier No. HYW004544500MG
1,10-Phenanthroline, also known as o-Phenanthroline, is a potent metal chelator and MMP inhibitor. It has been shown to prevent the induction of chromosomal aberrations in streptozotocin-treated cells. This compound forms a characteristic red chelate with Fe2+, exhibiting maximum absorption at 510 nm.
  • Acts as a metal chelator
  • Prevents chromosomal aberrations in streptozotocin-treated cells
  • Forms a red chelate with Fe2+ with maximum absorption at 510 nm
  • Functions as an MMP inhibitor
  • Appears as a white to off-white solid